Assay Details
Target Gene Details
Entrez Gene ID: | 74610 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 8 |
Gene Aliases: |
4833412N02Rik, AA409895 |
Location: |
Chr.5:24393681-24409947 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abcb8 | NM_029020.2 | 12 | 1771 | NP_083296.2 |
AF213391.1 | 1 | 38 | AAF31430.1 | |
AK014319.1 | 12 | 1502 | BAB29270.1 | |
AK030624.1 | 12 | 1498 | BAC27052.1 | |
AK038760.1 | 12 | 1499 | ||
AK049152.1 | 12 | 1515 | BAC33571.1 | |
AK076315.1 | 12 | 1522 | BAC36297.1 | |
AK166704.1 | 12 | 1505 | BAE38958.1 | |
AK170334.1 | ||||
AK189429.1 | 2 | 112 | ||
AK196341.1 | 2 | 177 | ||
AK205793.1 | 2 | 125 | ||
BC015301.1 | 12 | 1771 | AAH15301.1 |
More Information
Set Membership: |
Intragenic Exonic |