Assay Details
Target Gene Details
Entrez Gene ID: | 319974 |
Gene Name: | autism susceptibility candidate 2 |
Gene Aliases: |
2700063G02Rik, A730011F23Rik, D830032G16Rik |
Location: |
Chr.5:131437682-132542343 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Auts2 | NM_177047.3 | NP_796021.2 | ||
AK052418.1 | BAC34980.1 | |||
AK085946.1 | ||||
BC151031.1 | ||||
BC151047.1 |