Assay Details
Target Gene Details
Entrez Gene ID: | 50500 |
Gene Name: | tocopherol (alpha) transfer protein |
Gene Aliases: |
alpha-TTP |
Location: |
Chr.4:20008089-20030785 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttpa | NM_015767.3 | 4 | 619 | NP_056582.1 |
AF218416.1 | 4 | 607 | AAF25956.1 | |
AK004882.1 | 4 | 600 | BAB23640.1 | |
AK050374.1 | 4 | 620 | BAC34218.1 | |
BC026882.1 | 4 | 506 | AAH26882.1 | |
BC099868.1 | 4 | 618 | AAH99868.1 | |
BC099869.1 | 4 | 620 | AAH99869.1 | |
BC099870.1 | 4 | 620 | AAH99870.1 | |
BC099871.1 | 4 | 619 | AAH99871.1 |
More Information
Set Membership: |
Intragenic Exonic |