Assay Details
Target Gene Details
Entrez Gene ID: | 242418 |
Gene Name: | DDB1 and CUL4 associated factor 10 |
Gene Aliases: |
AA959934, Wdr32 |
Location: |
Chr.4:45341735-45379732 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dcaf10 | NM_153167.2 | NP_694807.2 | ||
AK045241.1 | BAC32276.1 | |||
AK076417.1 | BAC36330.1 | |||
AK083428.1 | BAC38912.1 | |||
AK157345.1 | ||||
AK170534.1 | ||||
AK194080.1 | ||||
AK213618.1 | ||||
BC027317.1 | AAH27317.1 | |||
BC085101.1 | AAH85101.1 | |||
CN533345.1 |