Assay Details
Target Gene Details
Entrez Gene ID: | 54170 |
Gene Name: | Ras-related GTP binding C |
Gene Aliases: |
AU041672, Gtr2, RAGC, TIB929, YGR163W |
Location: |
Chr.4:123917433-123936997 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rragc | NM_017475.2 | 3 | 603 | NP_059503.2 |
AB017616.1 | 3 | 526 | BAA75671.1 | |
AK150792.1 | 3 | 557 | ||
AK159355.1 | 3 | 536 | BAE35013.1 | |
AK159434.1 | 3 | 558 | ||
AK166658.1 | 3 | 549 | BAE38923.1 | |
BC005417.1 | 3 | 551 | AAH05417.1 | |
BC071245.1 | 3 | 951 | AAH71245.1 |
More Information
Set Membership: |
Intragenic Exonic |