Assay Details
Target Gene Details
Entrez Gene ID: | 76376 |
Gene Name: | solute carrier family 24 (sodium/potassium/calcium exchanger), member 2 |
Gene Aliases: |
2810021B17Rik, 6330417K15Rik, AI847460, Nckx2 |
Location: |
Chr.4:86983124-87230554 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc24a2 | NM_001110240.1 | NP_001103710.1 | ||
NM_172426.2 | NP_766014.1 | |||
AK083127.1 | BAC38773.1 | |||
BC115867.1 | ||||
BC116637.1 |
More Information
Set Membership: |
Intragenic Non-exonic |