Assay Details
Target Gene Details
Entrez Gene ID: | 242285 |
Gene Name: | short chain dehydrogenase/reductase family 16C, member 5 |
Gene Aliases: |
Rdhe2, Scdr9 |
Location: |
Chr.4:3995942-4019663 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sdr16c5 | NM_181989.1 | NP_871789.1 | ||
AY277588.1 | AAP41074.1 | |||
BC104134.1 | AAI04135.1 | |||
BC104135.1 | AAI04136.1 |