Assay Details
Target Gene Details
Entrez Gene ID: | 14211 |
Gene Name: | structural maintenance of chromosomes 2 |
Gene Aliases: |
5730502P04Rik, AI255214, AW545314, CAP-E, CAPE, Fin16, SMC-2, Smc2l1 |
Location: |
Chr.4:52439221-52488365 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smc2 | NM_001301412.1 | 16 | 2313 | NP_001288341.1 |
NM_008017.4 | 16 | 2237 | NP_032043.3 | |
AJ534939.1 | 15 | 2068 | CAD59182.1 | |
AK028233.1 | 16 | 2280 | ||
AK145341.1 | 16 | 2267 | BAE26376.1 | |
AK152090.1 | 16 | 2265 | ||
BC071232.1 | AAH71232.1 | |||
BC094380.1 | 16 | 2209 | AAH94380.1 |