Assay Details
Target Gene Details
Entrez Gene ID: | 14745 |
Gene Name: | lysophosphatidic acid receptor 1 |
Gene Aliases: |
AI326300, Edg2, Gpcr26, Kdt2, lpA1, vzg-1 |
Location: |
Chr.4:58435252-58554027 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lpar1 | NM_010336.2 | NP_034466.2 | ||
NM_172989.1 | NP_766577.1 | |||
AK030330.1 | BAC26905.1 | |||
BC025425.1 | AAH25425.1 | |||
BY716867.1 | ||||
CJ152526.1 | ||||
CJ175997.1 | ||||
U70622.1 | AAC52923.1 |
More Information
Set Membership: |
Intragenic Non-exonic |