Assay Details
Target Gene Details
Entrez Gene ID: | 18198 |
Gene Name: | muscle, skeletal, receptor tyrosine kinase |
Gene Aliases: |
Mdk4, Mlk, Nsk1, Nsk2, Nsk3 |
Location: |
Chr.4:58285640-58377848 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Musk | NM_001037127.2 | 13 | 1739 | NP_001032204.2 |
NM_001037128.1 | 13 | 1694 | NP_001032205.1 | |
NM_001037129.1 | 12 | 1670 | NP_001032206.1 | |
NM_001037130.1 | 11 | 1640 | NP_001032207.1 | |
NM_010944.2 | 12 | 1664 | NP_035074.2 | |
AB691772.1 | 11 | 1490 | ||
AY955296.1 | 13 | 1589 | AAY40318.1 | |
AY955297.1 | 13 | 1544 | AAY40319.1 | |
AY955298.1 | 12 | 1520 | AAY40320.1 | |
AY956403.1 | 12 | 1514 | AAY40321.1 | |
AY956404.1 | 11 | 1490 | AAY40322.1 | |
BC100342.1 | 12 | 1594 | AAI00343.1 | |
U37708.1 | 11 | 1638 | AAA79203.1 | |
U37709.1 | 12 | 1662 | AAA79204.1 | |
X86444.1 | 11 | 1568 | CAA60165.1 |
More Information
Set Membership: |
Intragenic Exonic |