Assay Details
Target Gene Details
Entrez Gene ID: | 230101 |
Gene Name: | glucosidase beta 2 |
Gene Aliases: |
F630034E04 |
Location: |
Chr.4:43566928-43578864 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gba2 | NM_172692.3 | NP_766280.2 | ||
AK089192.1 | BAC40785.1 | |||
AK158174.1 | ||||
AK173213.1 | BAD32491.1 | |||
AK180481.1 | ||||
AK188452.1 | ||||
BC056935.1 | AAH56935.1 |
More Information
Set Membership: |
Intragenic Non-exonic |