Assay Details
Target Gene Details
Entrez Gene ID: | 13841 |
Gene Name: | Eph receptor A7 |
Gene Aliases: |
Cek11, Ebk, Ehk3, Hek11, Mdk1 |
Location: |
Chr.4:28813044-28967511 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epha7 | NM_001122889.1 | 7 | 1742 | NP_001116361.1 |
NM_001290434.1 | 7 | 1756 | NP_001277363.1 | |
NM_010141.4 | 7 | 1756 | NP_034271.3 | |
AK030480.1 | 7 | 1723 | BAC26982.1 | |
AK033903.1 | 7 | 1757 | BAC28509.1 | |
AK049848.1 | 7 | 1723 | ||
BC026153.1 | 7 | 1648 | AAH26153.1 | |
X79082.1 | 7 | 1735 | CAA55687.1 | |
X79083.1 | 7 | 1735 | CAA55688.1 | |
X79084.1 | 7 | 1735 | CAA55689.1 | |
X81466.1 | 3 | 213 | CAA57224.1 |
More Information
Set Membership: |
Intragenic Exonic |