Assay Details
Target Gene Details
Entrez Gene ID: | 13841 |
Gene Name: | Eph receptor A7 |
Gene Aliases: |
Cek11, Ebk, Ehk3, Hek11, Mdk1 |
Location: |
Chr.4:28813044-28967511 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epha7 | NM_001122889.1 | NP_001116361.1 | ||
NM_001290434.1 | NP_001277363.1 | |||
NM_010141.4 | NP_034271.3 | |||
AK030480.1 | BAC26982.1 | |||
AK032973.1 | BAE43283.1 | |||
AK033903.1 | BAC28509.1 | |||
AK049848.1 | ||||
BC026153.1 | AAH26153.1 | |||
CF729611.1 | ||||
X79082.1 | CAA55687.1 | |||
X79083.1 | CAA55688.1 | |||
X79084.1 | CAA55689.1 |
More Information
Set Membership: |
Intragenic Non-exonic |