Assay Details
Target Gene Details
Entrez Gene ID: | 13841 |
Gene Name: | Eph receptor A7 |
Gene Aliases: |
Cek11, Ebk, Ehk3, Hek11, Mdk1 |
Location: |
Chr.4:28813044-28967511 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epha7 | NM_001290434.1 | 15 | 2870 | NP_001277363.1 |
NM_010141.4 | 15 | 2882 | NP_034271.3 | |
AK030480.1 | 15 | 2849 | BAC26982.1 | |
BC026153.1 | 15 | 2762 | AAH26153.1 | |
X79082.1 | 15 | 2861 | CAA55687.1 | |
X79084.1 | CAA55689.1 | |||
X81466.1 | 11 | 1339 | CAA57224.1 |
More Information
Set Membership: |
Intragenic Exonic |