Assay Details
Target Gene Details
Entrez Gene ID: | 72479 |
Gene Name: | hydroxysteroid dehydrogenase like 2 |
Gene Aliases: |
2610207I16Rik |
Location: |
Chr.4:59580458-59618694 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hsdl2 | NM_001301367.1 | 4 | 530 | NP_001288296.1 |
NM_024255.3 | 3 | 432 | NP_077217.2 | |
AJ293845.1 | 4 | 453 | CAC34587.1 | |
AK011910.1 | 3 | 390 | BAB27910.1 | |
AK085899.1 | 3 | 401 | BAC39563.1 | |
AK136773.1 | 3 | 399 | BAE23126.1 | |
AK144948.1 | 3 | 348 | ||
AK145230.1 | 3 | 292 | BAE26313.1 | |
AK202946.1 | 2 | 147 | ||
AK205169.1 | 2 | 147 | ||
AK211849.1 | 2 | 147 | ||
AK213102.1 | 2 | 139 | ||
AK215396.1 | 2 | 147 | ||
AY725196.1 | 3 | 289 | AAU11505.1 | |
BC002211.1 | 3 | 305 | AAH02211.1 |
More Information
Set Membership: |
Intragenic Exonic |