Assay Details
Target Gene Details
Entrez Gene ID: | 72479 |
Gene Name: | hydroxysteroid dehydrogenase like 2 |
Gene Aliases: |
2610207I16Rik |
Location: |
Chr.4:59580458-59618694 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hsdl2 | NM_001301367.1 | 3 | 440 | NP_001288296.1 |
NM_024255.3 | 2 | 342 | NP_077217.2 | |
AJ293845.1 | 3 | 363 | CAC34587.1 | |
AK011910.1 | 2 | 300 | BAB27910.1 | |
AK085899.1 | 2 | 311 | BAC39563.1 | |
AK136773.1 | 2 | 309 | BAE23126.1 | |
AK144948.1 | 2 | 258 | ||
AK145230.1 | 2 | 202 | BAE26313.1 | |
AK202946.1 | 1 | 57 | ||
AK205169.1 | 1 | 57 | ||
AK211849.1 | 1 | 57 | ||
AK213102.1 | ||||
AK215396.1 | 1 | 57 | ||
AY725196.1 | 2 | 199 | AAU11505.1 | |
BC002211.1 | 2 | 215 | AAH02211.1 | |
BY327952.1 |
More Information
Set Membership: |
Intragenic Exonic |