Assay Details
Target Gene Details
Entrez Gene ID: | 17025 |
Gene Name: | aminolevulinate, delta-, dehydratase |
Gene Aliases: |
ALADH, Lv |
Location: |
Chr.4:62505984-62520063 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Alad | NM_001276446.1 | NP_001263375.1 | ||
NM_008525.4 | NP_032551.3 | |||
AK002300.1 | BAB21998.1 | |||
AK032908.1 | BAC28080.1 | |||
AK167673.1 | BAE39722.1 | |||
AK168119.1 | BAE40090.1 | |||
AK168132.1 | BAE40101.1 | |||
AK178951.1 | ||||
BC018236.1 | AAH18236.1 | |||
BC055930.1 | AAH55930.1 | |||
CB524263.1 | ||||
U89407.1 | AAC36514.1 | |||
X13752.1 | CAA32015.1 |
More Information
Set Membership: |
Intragenic Non-exonic |