Assay Details
Target Gene Details
Entrez Gene ID: | 230787 |
Gene Name: | thymocyte selection associated family member 2 |
Gene Aliases: |
ICB-1 |
Location: |
Chr.4:132781837-132796364 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Themis2 | NM_001033308.2 | NP_001028480.1 | ||
AK137465.1 | ||||
AK149775.1 | ||||
AK150471.1 | BAE29588.1 | |||
AK153536.1 | BAE32073.1 | |||
AK155536.1 | BAE33313.1 | |||
AK171009.1 | BAE42180.1 | |||
AK171858.1 | ||||
BC013712.1 | AAH13712.1 | |||
BC151122.1 | ||||
BC151126.1 |