Assay Details
Target Gene Details
Entrez Gene ID: | 12017 |
Gene Name: | BCL2-associated athanogene 1 |
Gene Aliases: |
BAG-1, Rap46 |
Location: |
Chr.4:40936398-40948294 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bag1 | NM_001171739.1 | NP_001165210.1 | ||
NM_009736.3 | NP_033866.4 | |||
AF022223.1 | AAC34259.1 | |||
AK009149.1 | BAB26106.1 | |||
AK013611.1 | BAB28930.1 | |||
AK154469.1 | ||||
AK171094.1 | ||||
AK188292.1 | ||||
BC003722.1 | ||||
BC069918.1 | ||||
BC093509.1 | ||||
BQ031093.1 |