Assay Details
Target Gene Details
Entrez Gene ID: | 230235 |
Gene Name: | ferric-chelate reductase 1 like |
Gene Aliases: |
6430704M03Rik |
Location: |
Chr.4:56960136-56990391 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Frrs1l | NM_001142965.1 | 3 | 510 | NP_001136437.1 |
AK038461.1 | 3 | 214 | BAC30008.1 | |
AK078331.1 | 2 | 173 | BAC37221.1 | |
BC132345.1 | 3 | 537 |