Assay Details
Target Gene Details
Entrez Gene ID: | 97064 |
Gene Name: | WW domain containing transcription regulator 1 |
Gene Aliases: |
2310058J06Rik, 2610021I22Rik, C78399, Taz |
Location: |
Chr.3:57455644-57575910 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wwtr1 | NM_001168281.1 | NP_001161753.1 | ||
NM_133784.3 | NP_598545.2 | |||
AJ299430.1 | CAC17733.1 | |||
AK009983.1 | ||||
AK011504.1 | ||||
AK142299.1 | BAE25019.1 | |||
AK142720.1 | BAE25174.1 | |||
BC004640.1 | AAH04640.1 | |||
BC014727.1 | AAH14727.1 |
More Information
Set Membership: |
Intragenic Non-exonic |