Assay Details
Target Gene Details
Entrez Gene ID: | 70093 |
Gene Name: | ubiquitin-conjugating enzyme E2Q (putative) 1 |
Gene Aliases: |
1110002C01Rik, 2310012M18Rik, AI854014, Gtap, NICE-5, PRO3094, Ube2q |
Location: |
Chr.3:89773609-89783997 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ube2q1 | NM_027315.4 | NP_081591.3 | ||
AK009324.1 | BAB26217.2 | |||
AK146707.1 | BAE27373.1 | |||
AK177704.1 | ||||
AK214106.1 | ||||
AY112699.1 | AAM60815.1 | |||
AY945937.1 | AAY23286.1 | |||
BC030044.1 | AAH30044.2 | |||
BC060966.1 | AAH60966.1 | |||
BC082275.1 | AAH82275.1 |
More Information
Set Membership: |
Intragenic Non-exonic |