Assay Details
Target Gene Details
Entrez Gene ID: | 18741 |
Gene Name: | paired-like homeodomain transcription factor 2 |
Gene Aliases: |
9430085M16Rik, Brx1, Brx1b, Munc30, Otlx2, Ptx2, Rieg |
Location: |
Chr.3:129199878-129219594 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pitx2 | NM_001042502.2 | 2 | 915 | NP_001035967.1 |
NM_001042504.2 | 3 | 406 | NP_001035969.1 | |
NM_001286942.1 | 4 | 505 | NP_001273871.1 | |
NM_001287048.1 | 2 | 915 | NP_001273977.1 | |
NM_011098.4 | 4 | 544 | NP_035228.2 | |
AB006320.1 | 2 | 468 | BAA75247.1 | |
AB006321.1 | 4 | 504 | BAA75248.1 | |
AF048723.1 | 3 | 343 | AAC40086.1 | |
AF048724.1 | 2 | 876 | AAC40087.1 | |
AF201091.1 | 2 | 242 | AAF44618.1 | |
AK034846.1 | 1 | 1774 | ||
AK035083.1 | 2 | 573 | BAC28939.1 | |
AM940438.1 | 3 | 210 | ||
AM940439.1 | 2 | 168 | ||
BC075660.1 | 2 | 580 | AAH75660.1 | |
U70132.1 | 1 | 65 | AAB38864.1 | |
U80010.1 | 4 | 479 | AAC53119.1 | |
U80011.1 | 3 | 359 | AAC53120.1 | |
U80036.1 | 3 | 377 | AAB38505.1 |
More Information
Set Membership: |
Intragenic Exonic |