Assay Details
Target Gene Details
Entrez Gene ID: | 66868 |
Gene Name: | major facilitator superfamily domain containing 1 |
Gene Aliases: |
1200003O06Rik |
Location: |
Chr.3:67582768-67604231 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mfsd1 | NM_025813.3 | 3 | 314 | NP_080089.1 |
AK004586.1 | 3 | 315 | BAB23391.1 | |
AK032688.1 | 3 | 283 | ||
AK075859.1 | 3 | 344 | BAC36010.1 | |
AK135235.1 | 3 | 315 | BAE22467.1 | |
AK136108.1 | 3 | 306 | BAE22824.1 | |
AK152444.1 | 3 | 336 | BAE31224.1 | |
AK159554.1 | 3 | 342 | BAE35179.1 | |
AK166645.1 | ||||
AK183168.1 | 2 | 79 | ||
AK218750.1 | ||||
BC024891.1 | 3 | 296 | AAH24891.1 |