Assay Details
Target Gene Details
Entrez Gene ID: | 210529 |
Gene Name: | methyltransferase like 14 |
Gene Aliases: |
G430022H21Rik, mKIAA1627 |
Location: |
Chr.3:123368295-123385990 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mettl14 | NM_201638.2 | NP_964000.2 | ||
AK083169.1 | ||||
AK089949.1 | ||||
AK146881.1 | BAE27502.1 | |||
AK153810.1 | ||||
BC052204.1 | AAH52204.1 |
More Information
Set Membership: |
Intragenic Non-exonic |