Assay Details
Target Gene Details
Entrez Gene ID: | 269437 |
Gene Name: | phospholipase C, eta 1 |
Gene Aliases: |
BC042549, PLCeta1, Plcl3 |
Location: |
Chr.3:63696232-63899677 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plch1 | NM_001177732.1 | 14 | 1816 | NP_001171203.1 |
NM_001177733.1 | 14 | 1816 | NP_001171204.1 | |
NM_183191.3 | 14 | 1816 | NP_899014.2 | |
AK173095.1 | 6 | 699 | BAD32373.1 | |
AY691172.1 | 14 | 1816 | AAW22609.1 | |
AY691173.1 | 14 | 1816 | AAW22610.1 | |
AY691174.1 | 14 | 1816 | AAW22611.1 | |
BC042549.1 | 2 | 145 | AAH42549.1 | |
BC052372.1 | 2 | 145 | AAH52372.1 | |
BC055005.1 | 14 | 1890 | AAH55005.1 |
More Information
Set Membership: |
Intragenic Exonic |