Assay Details
Target Gene Details
Entrez Gene ID: | 78523 |
Gene Name: | mitochondrial ribosomal protein L9 |
Gene Aliases: |
8030480E20Rik, AA409733, C330013D18Rik, L9mt |
Location: |
Chr.3:94443272-94448708 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mrpl9 | NM_030116.2 | 2 | 272 | NP_084392.1 |
AB049637.1 | 2 | 232 | BAB40842.1 | |
AK021196.1 | 2 | 291 | ||
AK040055.1 | 2 | 273 | BAC30503.1 | |
AK046099.1 | 2 | 272 | BAC32604.1 | |
AK161563.1 | 2 | 276 | ||
AK162023.1 | 2 | 272 | BAE36685.1 | |
BC116385.1 | 2 | 272 | ||
BC116386.1 | 2 | 272 |
More Information
Set Membership: |
Intragenic Exonic |