Assay Details
Target Gene Details
Entrez Gene ID: | 76089 |
Gene Name: | Rap guanine nucleotide exchange factor (GEF) 2 |
Gene Aliases: |
5830453M24Rik, CNRasGEF, Pdzgef1, RA-GEF-1, mKIAA0313, nRapGEP |
Location: |
Chr.3:79062516-79287244 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rapgef2 | NM_001099624.2 | 12 | 1781 | NP_001093094.2 |
AB093228.1 | 5 | 664 | BAC41412.1 | |
AK147368.1 | 12 | 1708 | ||
BC067056.1 | 12 | 1781 | AAH67056.1 |
More Information
Set Membership: |
Intragenic Exonic |