Assay Details
Target Gene Details
Entrez Gene ID: | 229317 |
Gene Name: | eukaryotic translation initiation factor 2A |
Gene Aliases: |
D030048D22, D3Ertd194e |
Location: |
Chr.3:58525821-58557501 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif2a | NM_001005509.2 | NP_001005509.1 | ||
AK044865.1 | ||||
AK050971.1 | ||||
AK078519.1 | BAC37320.1 | |||
AK135515.1 | 3 | 1989 | ||
AK145275.1 | ||||
AK161904.1 | 9 | 771 | ||
BC030447.1 | ||||
BC082557.1 | AAH82557.1 |
More Information
Set Membership: |
Intragenic Non-exonic |