Assay Details
Target Gene Details
Entrez Gene ID: | 20585 |
Gene Name: | helicase-like transcription factor |
Gene Aliases: |
AF010600, BC057116, P113, Smarca3, Snf2l3 |
Location: |
Chr.3:20057811-20118491 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hltf | NM_009210.3 | 7 | 926 | NP_033236.2 |
NR_105046.1 | 7 | 926 | ||
NR_105047.1 | 7 | 963 | ||
AB221636.1 | 7 | 749 | ||
AF010600.1 | 7 | 926 | AAB63915.1 | |
AF165911.1 | 7 | 905 | ||
AK141235.1 | 7 | 919 | BAE24607.1 | |
AK142961.1 | 7 | 883 | BAE25232.1 | |
AK149897.1 | 7 | 964 | ||
AK166911.1 | 7 | 881 | ||
BC057116.1 | 7 | 895 | AAH57116.1 | |
BC059240.1 | 7 | 895 | AAH59240.1 |