Assay Details
Target Gene Details
Entrez Gene ID: | 12630 |
Gene Name: | complement component factor i |
Gene Aliases: |
- |
Location: |
Chr.3:129835889-129875329 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cfi | NM_007686.2 | NP_031712.2 | ||
BC150751.1 | ||||
BG072327.1 | ||||
U47810.1 | AAB00438.1 |
More Information
Set Membership: |
Intragenic Non-exonic |