Assay Details
Target Gene Details
Entrez Gene ID: | 72007 |
Gene Name: | fibronectin type III domain containing 3B |
Gene Aliases: |
1600019O04Rik, AW550168, Fad104, mKIAA4164 |
Location: |
Chr.3:27416161-27711662 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fndc3b | NM_173182.2 | 19 | 2332 | NP_775274.2 |
AB098596.1 | 19 | 2270 | BAC53726.1 | |
AK147346.1 | 20 | 2343 | ||
AK220297.1 | 19 | 2332 | BAD90222.1 | |
AY234860.1 | 1 | 74 | AAO89277.1 | |
BC067389.1 | 19 | 2338 | AAH67389.1 | |
DQ192037.1 | 18 | 2238 | ABA82150.1 |
More Information
Set Membership: |
Intragenic Exonic |