Assay Details
Target Gene Details
Entrez Gene ID: | 72007 |
Gene Name: | fibronectin type III domain containing 3B |
Gene Aliases: |
1600019O04Rik, AW550168, Fad104, mKIAA4164 |
Location: |
Chr.3:27416161-27711662 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fndc3b | NM_173182.2 | NP_775274.2 | ||
AB098596.1 | BAC53726.1 | |||
AK147346.1 | ||||
AK220297.1 | BAD90222.1 | |||
BC067389.1 | AAH67389.1 | |||
DQ192037.1 | ABA82150.1 |
More Information
Set Membership: |
Intragenic Non-exonic |