Assay Details
Target Gene Details
Entrez Gene ID: | 12350 |
Gene Name: | carbonic anhydrase 3 |
Gene Aliases: |
BB219044, Ca3, Car-3 |
Location: |
Chr.3:14863490-14872536 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Car3 | NM_007606.3 | NP_031632.2 | ||
AK002349.1 | ||||
AK003671.1 | ||||
BC011129.1 | AAH11129.1 | |||
M27796.1 | AAA37355.1 |