Assay Details
Target Gene Details
Entrez Gene ID: | 229600 |
Gene Name: | cDNA sequence BC028528 |
Gene Aliases: |
L259 |
Location: |
Chr.3:95882352-95892949 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BC028528 | NM_153513.2 | NP_705733.2 | ||
AF534879.1 | AAN03482.1 | |||
BC028528.1 | AAH28528.1 |