Assay Details
Target Gene Details
Entrez Gene ID: | 15168 |
Gene Name: | hyperpolarization-activated, cyclic nucleotide-gated K+ 3 |
Gene Aliases: |
BCNG-4, Bcng4, Hac3, Hcn4 |
Location: |
Chr.3:89146775-89160228 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hcn3 | NM_008227.1 | NP_032253.1 | ||
AJ225124.1 | CAA12408.1 | |||
AK032225.1 | BAC27769.1 | |||
BC039156.1 | AAH39156.1 | |||
BC138319.1 | ||||
BC138321.1 |
More Information
Set Membership: |
Intragenic Non-exonic |