Assay Details
Target Gene Details
Entrez Gene ID: | 100042889 |
Gene Name: | RIKEN cDNA C920021L13 gene |
Gene Aliases: |
- |
Location: |
Chr.3:95871522-95889093 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C920021L13Rik | NR_040446.1 | |||
AK044447.1 | ||||
AK160363.1 |
Target Gene Details
Entrez Gene ID: | 229599 |
Gene Name: | circadian associated repressor of transcription |
Gene Aliases: |
Chrono, Gm129 |
Location: |
Chr.3:95878500-95882421 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ciart | NM_001033302.2 | NP_001028474.1 | ||
AK156084.1 | BAE33580.1 | |||
AK164001.1 | BAE37582.1 | |||
BC132471.1 | ||||
BC138510.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |