Assay Details
Target Gene Details
Entrez Gene ID: | 56811 |
Gene Name: | dickkopf homolog 2 (Xenopus laevis) |
Gene Aliases: |
- |
Location: |
Chr.3:132085242-132180304 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dkk2 | NM_020265.4 | 2 | 1076 | NP_064661.2 |
AJ243963.2 | 2 | 476 | CAB60110.1 | |
AK028225.1 | 2 | 361 | BAC25824.1 | |
AK031749.1 | 2 | 1077 | BAC27536.1 | |
BC096448.1 | 2 | 1067 | AAH96448.1 |