Assay Details
Target Gene Details
Entrez Gene ID: | 229499 |
Gene Name: | Fc receptor-like 1 |
Gene Aliases: |
A230020G22Rik, Bxmas1, Bxmh1, Bxmh1b, Fcrh1, Fcrh1l, Fcrh1s, Ifgp1 |
Location: |
Chr.3:87376333-87402934 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fcrl1 | NM_001136236.1 | NP_001129708.1 | ||
NM_153090.2 | NP_694730.2 | |||
NM_178165.4 | NP_835459.1 | |||
AK041750.1 |
More Information
Set Membership: |
Intragenic Non-exonic |