Assay Details
Target Gene Details
Entrez Gene ID: | 213262 |
Gene Name: | follistatin-like 5 |
Gene Aliases: |
9130207J01Rik |
Location: |
Chr.3:76074275-76710010 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fstl5 | NM_001253719.1 | NP_001240648.1 | ||
NM_178673.4 | NP_848788.2 | |||
AF374460.1 | AAQ02778.1 | |||
AK033652.1 | BAC28408.1 | |||
AK046070.1 | BAC32590.1 | |||
AK081152.1 | BAC38146.1 | |||
AK122482.1 | BAC65764.1 | |||
AK138388.1 | BAE23641.1 | |||
BC075640.1 | AAH75640.1 |
More Information
Set Membership: |
Intragenic Non-exonic |