Assay Details
Target Gene Details
Entrez Gene ID: | 73668 |
Gene Name: | tetratricopeptide repeat domain 21B |
Gene Aliases: |
2410066K11Rik, Thm1, aln, mKIAA1992 |
Location: |
Chr.2:66184327-66256675 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc21b | NM_001047604.2 | 24 | 3307 | NP_001041069.1 |
NM_001290669.1 | 24 | 3307 | NP_001277598.1 | |
AK132240.1 | 10 | 1226 | BAE21052.1 | |
AK173320.1 | 2 | 247 | BAD32598.1 | |
DQ011164.1 | 24 | 3307 |