Assay Details
Target Gene Details
Entrez Gene ID: | 66715 |
Gene Name: | HEN1 methyltransferase homolog 1 (Arabidopsis) |
Gene Aliases: |
4921515J06Rik, Hen1, mHEN1 |
Location: |
Chr.3:108938542-108960776 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Henmt1 | NM_001078646.1 | 7 | 831 | NP_001072114.1 |
NM_025723.2 | 7 | 1146 | NP_079999.2 | |
AK014900.1 | 7 | 1112 | ||
AK038994.1 | 7 | 832 | BAC30196.1 | |
AK085587.1 | 7 | 1146 | BAC39480.1 | |
BC099408.1 | 6 | 812 | AAH99408.1 | |
BC128494.1 | 5 | 627 | ||
BC128495.1 | 5 | 627 |