Assay Details
Target Gene Details
Entrez Gene ID: | 75812 |
Gene Name: | taspase, threonine aspartase 1 |
Gene Aliases: |
4930485D02Rik, AW986064 |
Location: |
Chr.2:139833479-140067666 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tasp1 | NM_001159640.2 | NP_001153112.1 | ||
NM_001159641.2 | NP_001153113.1 | |||
NM_001289611.1 | NP_001276540.1 | |||
NM_175225.5 | NP_780434.1 | |||
NR_110350.1 | ||||
NR_110351.1 | ||||
NR_110352.1 | ||||
AK015624.1 | ||||
AK036512.1 | BAC29457.1 | |||
AK132709.1 | ||||
AK146320.1 | BAE27075.1 | |||
AK160983.1 | ||||
BB665115.1 | ||||
BC024597.1 | AAH24597.1 | |||
BY055918.1 |
More Information
Set Membership: |
Intragenic Non-exonic |