Assay Details
Target Gene Details
Entrez Gene ID: | 76080 |
Gene Name: | tocopherol (alpha) transfer protein-like |
Gene Aliases: |
3110080A02Rik, 5830472M02Rik |
Location: |
Chr.2:163602281-163619013 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttpal | NM_029512.2 | NP_083788.2 | ||
NM_181734.3 | NP_859423.2 | |||
AK018052.1 | ||||
AK033600.1 | BAC28381.1 | |||
AK039450.1 | BAC30352.1 | |||
AK043791.1 | BAC31654.1 | |||
AK081680.1 | BAC38289.1 | |||
AK156641.1 | BAE33788.1 | |||
BC052389.1 | ||||
BY717985.1 | ||||
CK640285.1 |
More Information
Set Membership: |
Intragenic Non-exonic |