Assay Details
Target Gene Details
Entrez Gene ID: | 114644 |
Gene Name: | solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3 |
Gene Aliases: |
NaDC-3, NaDC3, SDCT2 |
Location: |
Chr.2:165405295-165473225 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc13a3 | NM_054055.2 | NP_473396.1 | ||
AF306491.1 | AAK49898.1 | |||
AK085326.1 | ||||
AK087518.1 | ||||
AK128982.1 | ||||
AK138344.1 | BAE23629.1 | |||
AK157881.1 | ||||
AK165355.1 | ||||
BC026803.1 | AAH26803.1 |
More Information
Set Membership: |
Intragenic Non-exonic |