Assay Details
Target Gene Details
Entrez Gene ID: | 27176 |
Gene Name: | ribosomal protein L7A |
Gene Aliases: |
Surf3 |
Location: |
Chr.2:26910807-26913311 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rpl7a | NM_013721.3 | NP_038749.1 | ||
AK019449.1 | BAB31725.1 | |||
AK135447.1 | BAE22536.1 | |||
AK160044.1 | BAE35586.1 | |||
AK160621.1 | BAE35921.1 | |||
AK160622.1 | BAE35922.1 | |||
BC052339.1 | AAH52339.1 | |||
BC065176.1 | AAH65176.1 | |||
BC080663.1 | AAH80663.1 | |||
BC080669.1 | AAH80669.1 | |||
BC080712.1 | AAH80712.1 | |||
BC084678.1 | AAH84678.1 | |||
BC089624.1 | AAH89624.1 | |||
BC091731.1 | AAH91731.1 | |||
BC091769.1 | AAH91769.1 | |||
BC099389.1 | AAH99389.1 |
More Information
Set Membership: |
Intragenic Non-exonic |