Assay Details
Target Gene Details
Entrez Gene ID: | 241770 |
Gene Name: | regulating synaptic membrane exocytosis 4 |
Gene Aliases: |
6530406L02, Rim4 |
Location: |
Chr.2:163859756-163918990 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rims4 | NM_183023.1 | 6 | 637 | NP_898844.1 |
AY326955.1 | 6 | 637 | AAQ01682.1 | |
BC115475.1 | 6 | 636 |
More Information
Set Membership: |
Intragenic Exonic |