Assay Details
Target Gene Details
Entrez Gene ID: | 19261 |
Gene Name: | signal-regulatory protein alpha |
Gene Aliases: |
AI835480, Bit, CD172a, P84, Ptpns1, SHP-1, SHPS-1, SIRP |
Location: |
Chr.2:129592606-129632228 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sirpa | NM_001177647.2 | 3 | 933 | NP_001171118.1 |
NM_001291019.1 | 5 | 1587 | NP_001277948.1 | |
NM_001291020.1 | 6 | 1389 | NP_001277949.1 | |
NM_001291021.1 | 3 | 933 | NP_001277950.1 | |
NM_001291022.1 | 2 | 576 | NP_001277951.1 | |
NM_007547.4 | 5 | 1587 | NP_031573.2 | |
AB018194.1 | 5 | 1153 | BAA76555.1 | |
AF072543.1 | 5 | 1143 | AAC24886.1 | |
AF072544.1 | 3 | 489 | AAC24887.1 | |
AF332079.1 | 5 | 1153 | AAK56107.1 | |
AF332080.1 | 5 | 1153 | AAK56108.1 | |
AK159617.1 | 6 | 1396 | ||
AK189165.1 | 2 | 156 | ||
AK195466.1 | 2 | 152 | ||
BC062197.1 | 5 | 1178 | AAH62197.1 | |
CJ151068.1 | 2 | 177 | ||
CX236057.1 | 3 | 518 | ||
D85785.1 | 5 | 1437 | BAA20376.1 | |
D87967.1 | 5 | 1552 | BAA13520.1 | |
D87968.1 | 5 | 1552 | BAA13521.1 | |
U89694.1 | 5 | 1164 | AAB92591.1 | |
Y10349.1 | 5 | 1198 | CAA71375.1 |
More Information
Set Membership: |
Intragenic Exonic |