Assay Details
Target Gene Details
Entrez Gene ID: | 18798 |
Gene Name: | phospholipase C, beta 4 |
Gene Aliases: |
A930039J07Rik, AI854601, C230058B11Rik |
Location: |
Chr.2:135658437-136014602 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plcb4 | NM_013829.2 | 12 | 932 | NP_038857.1 |
AF332071.1 | 12 | 932 | AAK56099.1 | |
AF332072.1 | 12 | 932 | AAK56100.1 | |
AK148001.1 | 13 | 1118 | ||
BC061201.1 | 13 | 1042 | AAH61201.1 | |
BC089521.1 | 12 | 895 | AAH89521.1 | |
BC129883.1 | 12 | 927 |
More Information
Set Membership: |
Intragenic Exonic |