Assay Details
Target Gene Details
Entrez Gene ID: | 18573 |
Gene Name: | phosphodiesterase 1A, calmodulin-dependent |
Gene Aliases: |
AI987702, AW125737 |
Location: |
Chr.2:79834453-80196518 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde1a | NM_001009978.1 | 12 | 1263 | NP_001009978.1 |
NM_001009979.1 | 12 | 1263 | NP_001009979.1 | |
NM_001159582.1 | 14 | 1765 | NP_001153054.1 | |
NM_016744.4 | 13 | 1560 | NP_058024.2 | |
AF159298.1 | 12 | 1263 | AAF63857.1 | |
AK014810.1 | 12 | 1259 | ||
AK043647.1 | 13 | 1561 | BAC31606.1 | |
AK050004.1 | 14 | 1627 | ||
AY845863.1 | 13 | 1312 | ||
AY845864.1 | 12 | 1263 | AAW31879.1 | |
BC090628.1 | 13 | 1528 | AAH90628.1 | |
U56649.1 | 14 | 1496 | AAB03319.1 |
More Information
Set Membership: |
Intragenic Exonic |